missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR3B Antibody (2A8E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61747-0.025ml
This item is not returnable.
View return policy
Description
NMDAR3B Monoclonal specifically detects NMDAR3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| NMDAR3B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| glutamate [NMDA] receptor subunit 3B, glutamate receptor, ionotropic, N-methyl-D-aspartate 3B, NMDA receptor subunit 3B, NMDA type glutamate receptor subunit NR3B, NMDAR3B, N-methyl-D-aspartate receptor subtype 3B, NR3B | |
| Mouse | |
| 113 kDa | |
| 0.025 mL | |
| 116444 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 2A8E11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRIN3B | |
| Purified recombinant fragment of human GRIN3B (AA: 135-276) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction