missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDARA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95176-0.02ml
This item is not returnable.
View return policy
Description
NMDARA1 Polyclonal antibody specifically detects NMDARA1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NMDARA1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| glutamate [NMDA] receptor-associated protein 1, glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1(glutamate binding), glutamate receptor, NMDA subtype, glutamate-binding subunit, HNRGW, LFG1, NMDA receptor glutamate-binding subunit, NMDARA1MGC99687, Putative MAPK-activating protein PM02, TMBIM3, transmembrane BAX inhibitor motif containing 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GRINA (NP_000828.1). MSHEKSFLVSGDNYPPPNPGYPGGPQPPMPPYAQPPYPGAPYPQPPFQPSPYGQPGYPHGPSPYPQGGYPQGPYPQGGYPQGPYPQEGYPQGPYPQGGYP | |
| 0.02 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 2907 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction