missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME1-NME2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05576-20ul
This item is not returnable.
View return policy
Description
NME1-NME2 Polyclonal antibody specifically detects NME1-NME2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NME1-NME2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| NM23-LV, NME1-NME2 protein, NME1-NME2 readthrough transcript, NMELV | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-267 of human NME1-NME2 (NP_001018146.1). MANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQASEDLLKEHYVDLKDRPFFAGLVKYMHSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRTMANLERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVAMKFLRASEEHLKQHYIDLKDRPFFPGLVKYMNSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVKSAEKEISLWFKPEELVDYKSCAHDWVYE | |
| 20 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 654364 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction