missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94554-0.02ml
This item is not returnable.
View return policy
Description
NME4 Polyclonal antibody specifically detects NME4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NME4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| EC 2.7.4.6, NDP kinase, mitochondrial, NDPK-D, NM23D, nm23-H4mitochondrial, non-metastatic cells 4, protein expressed in | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human NME4 (NP_005000.1). MGGLFWRSALRGLRCGPRAPGPSLLVRHGSGGPSWTRERTLVAVKPDGVQRRLVGDVIQRFERRGFTLVGMKMLQAPESVLAEHYQDLRRKPFYPALIRYMSSGPVVAMVWEGYNVVRASRAMIGHTDSAEAAPGTIRGDFSVHISRNVIHASDSVEGAQREIQLWFQSSELVSWADGGQHSSIHPA | |
| 0.02 mL | |
| Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 4833 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction