missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME4 Antibody (OTI1A5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73005DL594
This item is not returnable.
View return policy
Description
NME4 Monoclonal antibody specifically detects NME4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NME4 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.7.4.6, NDP kinase, mitochondrial, NDPK-D, NM23D, nm23-H4mitochondrial, non-metastatic cells 4, protein expressed in | |
| Full length recombinant protein of human NME4 (NP_005000) produced in HEK293T cells. | |
| 0.1 mL | |
| Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 4833 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1A5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction