missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NME5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565153
This item is not returnable.
View return policy
Description
Immunogen sequence: FMFPEVIVEPI PIGQAAKDYL NLHIMPTLLE GLTELCKQKP ADPLIWLADW LLKNNPNKPK LCHHPI Highest antigen sequence identity to the following orthologs - mouse 79%, rat 85%.
Does not seem to have NDK kinase activity. Confers protection from cell death by Bax and alters the cellular levels of several antioxidant enzymes including Gpx5. May play a role in spermiogenesis by increasing the ability of late-stage spermatids to eliminate reactive oxygen species.
Specifications
| NME5 | |
| Polyclonal | |
| Unconjugated | |
| NME5 | |
| 1700019D05Rik; expressed in non-metastatic cells 5; inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; NM23H5; NM23-H5; nm23-M5; NME/NM23 family member 5; NME5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; RSPH23; testis-specific nm23 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56597 | |
| NME5 | |
| Recombinant Human NME5. Recombinant protein control fragment (Product #RP-96871). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction