missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93423-0.1ml
This item is not returnable.
View return policy
Description
NME6 Polyclonal antibody specifically detects NME6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NME6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| EC 2.7.4.6, Inhibitor of p53-induced apoptosis-alpha, IPIA-ALPHA, NDK 6, NDP kinase 6, NM23-H6, non-metastatic cells 6, protein expressed in (nucleoside-diphosphate kinase), nucleoside diphosphate kinase 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 9-194 of human NME6 (NP_005784.1). MASILRSPQALQLTLALIKPDAVAHPLILEAVHQQILSNKFLIVRMRELLWRKEDCQRFYREHEGRFFYQRLVEFMASGPIRAYILAHKDAIQLWRTLMGPTRVFRARHVAPDSIRGSFGLTDTRNTTHGSDSVVSASREIAAFFPDFSEQRWYEEEEPQLRCGPVCYSPEGGVHYVAGTGGLGPA | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 10201 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction