Learn More
Invitrogen™ NMI Monoclonal Antibody (2F3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549239
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human NMI shares 64% amino acid (aa) sequence identity with mouse NMI. Positive Control - WB: human placenta tissue, K562 whole cell, Hela whole cell, A431 whole cell, PC-3 whole cell, Caco-2 whole cell, HEK293 whole cell, mouse intestine tissue, rat liver tissue. IHC: human lung cancer tissue, human colon cancer tissue, human placenta tissue. ICC/IF: SiHa cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
Specifications
| NMI | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O35309, Q13287 | |
| NMI | |
| E.coli-derived human NMI recombinant protein (Position: E2-E307). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2F3 | |
| Unconjugated | |
| NMI | |
| N-mcy (and STAT) interactor; Nmi; N-myc (and STAT) interactor; N-myc and STAT interactor; N-myc interactor; N-myc-interactor | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 311021, 64685, 9111 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.