Learn More
NMI Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809982S
This item is not returnable.
View return policy
Description
NMI Monoclonal antibody specifically detects NMI in Human samples. It is validated for Western BlotSpecifications
| NMI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZSZ6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10194 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI2C1 | |
| Unconjugated | |
| TSHZ1 | |
| N-mcy (and STAT) interactor; Nmi; N-myc (and STAT) interactor; N-myc and STAT interactor; N-myc interactor; N-myc-interactor | |
| Human recombinant protein fragment corresponding to amino acids 29-176 of human TSHZ1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |