Learn More
NMI Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809982
Encompass_Preferred
 View more versions of this product
Product Code. 17532694
  • € 540.87 / 100µL
Description
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
Specifications
Specifications
NMI
Monoclonal
Unconjugated
NMI
N-myc and STAT interactor
Mouse
Affinity Chromatography
RUO
9111
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2C1
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13287
NMI
Human recombit protein fragment corresponding to amino acids 1-223 of human NMI produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b