missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMNAT-1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97227UV
This item is not returnable.
View return policy
Description
NMNAT-1 Polyclonal antibody specifically detects NMNAT-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NMNAT-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| EC 2.7.7.1, EC 2.7.7.18, NaMN adenylyltransferase 1, nicotinamide mononucleotide adenylyltransferase 1, nicotinamide nucleotide adenylyltransferase, nicotinamide nucleotide adenylyltransferase 1, Nicotinate-nucleotide adenylyltransferase 1, NMN adenylyltransferase 1, NMNATPNAT1, pyridine nucleotide adenylyltransferase 1 | |
| Produced in rabbits immunized with E. coli-derived Human NMNAT-1 fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 64802 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction