missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMNAT3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142781
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_835471.1; NP_001186976.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate with the same efficiency. Can use triazofurin monophosphate (TrMP) as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity, can use NAD (+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+). Protects against axonal degeneration following injury.
Specifications
| NMNAT3 | |
| Polyclonal | |
| Unconjugated | |
| NMNAT3 | |
| 4933408N02Rik; FKSG76; naMN adenylyltransferase 1; naMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; nicotinamide nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; nicotinamide-nucleotide adenylyltransferase 3; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3; NMNAT3; PNAT3; PNAT-3; pyridine nucleotide adenylyltransferase 3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 74080 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q99JR6 | |
| NMNAT3 | |
| Peptide with sequence C-KALGQGQSVKYLLPE. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction