missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMRAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99301-50ul
This item is not returnable.
View return policy
Description
NMRAL1 Polyclonal antibody specifically detects NMRAL1 in Human samples. It is validated for Western Blot
Specifications
| NMRAL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HSCARGFLJ25918, NmrA-like family domain containing 1, nmrA-like family domain-containing protein 1, SDR48A1, short chain dehydrogenase/reductase family 48A, member 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NMRAL1. | |
| 50 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 57407 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction