missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NMS 873

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
10mg
50mg
Description
NMS 873 is a potent p97 ATPase (VCP) allosteric inhibitor (IC50 = 30 nM). Selective over all other AAA ATPases, HSP90 and kinases tested (IC50 >10 μM). Activates the unfolded protein response (UPR) and modulates autophagosome maturation. Exhibits antiproliferative activity in cancer cells in vitro (IC50 values are 400 and 700 nM for HCT116 and HeLa cells respectively). Reduces p97-sensitization to trypsin digestion.
Specifications
Specifications
| Chemical Name or Material | 3-[3-(Cyclopentylthio)-5-[[[2-methyl-4'-(methylsulfonyl)[1,1'-biphenyl]-4-yl]oxy]methyl]-4H-1,2,4-triazol-4-yl]pyridine |
| CAS | 1418013-75-8 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C27H28N4O3S2 |
| Formula Weight | Observed MW: 520.67 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction