Learn More
NMT1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32168IR
Product Code. 18764202
  • € 681.00 / 0.10mL
Description
NMT1 Polyclonal antibody specifically detects NMT1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence