missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NMT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15644645
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15644645 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15644645 Supplier Invitrogen™ Supplier No. PA550716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NMT1 protein.

NMT1 is involved with Myristate, a rare 14-carbon saturated fatty acid, is cotranslationally attached by an amide linkage to the N-terminal glycine residue of cellular and viral proteins with diverse functions. N-myristoyltransferase (NMT EC 2.3.1.97) catalyzes the transfer of myristate from CoA to proteins. N-myristoylation appears to be irreversible and is required for full expression of the biologic activities of several N-myristoylated proteins, including the alpha subunit of the signal-transducing guanine nucleotide-binding protein (G protein) GO (GNAO1 MIM 139311).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Nmt1
Gene Accession No. P30419
Gene Alias alternative, short form NMT-S; AW536594; Glycylpeptide N-tetradecanoyltransferase 1; long form, NMT-L; myristoyl-CoA:protein N-myristoyltransferase 1; NMT; NMT 1; Nmt1; N-myristoyltransferase 1; Peptide N-myristoyltransferase 1; type I N-myristoyltransferase
Gene Symbols Nmt1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human N-myristoyltransferase 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4836
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.