missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMT2 Monoclonal Antibody (OTI1G3)

NMT2 Monoclonal Antibody (OTI1G3)
Brand: Invitrogen MA525836
This item is not returnable.
View return policy
Description
Specifications
| NMT2 | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60551 | |
| NMT2 | |
| Full length human recombinant protein of NMT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G3 | |
| Unconjugated | |
| NMT2 | |
| A930001K02Rik; AI605445; AU044698; fb40e03; Glycylpeptide N-tetradecanoyltransferase 2; glycylpeptide N-tetradecanoyltransferase 2 variant 3; glycylpeptide N-tetradecanoyltransferase 2 variant 4; hNMT-2; myristoyl-CoA:protein N-myristoyltransferase 2; NMT 2; Nmt2; NMT2 protein; N-myristoyltransferase 2; peptide N-myristoyltransferase 2; Type II N-myristoyltransferase; wu:fb40e03 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9397 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction