Learn More
Invitrogen™ NMU Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA579746
Product Code. 15935315
  • € 460.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat kidney tissue, mouse kidney tissue.
This gene encodes a member of the neuromedin family of neuropeptides. The encoded protein is a precursor that is proteolytically processed to generate a biologically active neuropeptide that plays a role in pain, stress, immune-mediated inflammatory diseases and feeding regulation. Increased expression of this gene was observed in renal, pancreatic and lung cancers. Alternative splicing results in multiple transcript variants encoding different isoforms. Some of these isoforms may undergo similar processing to generate the mature peptide.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NMU
Polyclonal
Unconjugated
NMU
Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; neuromedin U; neuromedin-U; Neuromedin-U-23; Neuromedin-U-25; Nmu; NmU 25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU
Rabbit
Antigen affinity chromatography
RUO
10874, 56183, 63887
-20°C
Lyophilized
Western Blot
500 μg/mL
PBS with 4mg trehalose and 0.05mg sodium azide
P12760, P48645, Q9QXK8
NMU
A synthetic peptide corresponding to a sequence of human NMU (FRVDEEFQSPFASQSRGYFLFRPRN).