missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NNMT Polyclonal Antibody
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
Specifications
Specifications
| Antigen | NNMT |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | NNMT |
| Gene Accession No. | P40261 |
| Gene Alias | Nicotinamide N-methyltransferase; NNMT |
| Gene Symbols | NNMT |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?