missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NNMT Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143207
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
Specifications
| NNMT | |
| Polyclonal | |
| Unconjugated | |
| NNMT | |
| Nicotinamide N-methyltransferase; NNMT | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4837 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P40261 | |
| NNMT | |
| Peptide with sequence PDLPTYCRALRN. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction