missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nNOS Antibody (60718), Alexa Fluor 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB2416AF647
This item is not returnable.
View return policy
Description
nNOS Monoclonal specifically detects nNOS in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| nNOS | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NOS1 | |
| Partial recombinant human nNOS protein (amino acids 218-1434) [UniProt P29475] | |
| 100 μL | |
| Asthma, Cancer, HIF Target Genes, Hypoxia, Immunology, Neuroscience, Neurotransmission | |
| 4842 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 60718 | |
| Western Blot | |
| bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction