missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ nNOS Monoclonal Antibody (GT5978)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546909
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.
Specifications
| nNOS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P29476, Q9Z0J4 | |
| NOS1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of Rat nNOS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT5978 | |
| Unconjugated | |
| NOS1 | |
| 2310005C01Rik; bNOS; bNOS1; Brain NOS; Constitutive NOS; IHPS1; NC-NOS; neuronal nitric oxide synthase; neuronal nitric oxide synthase NOS1; neuronal NOS; nitric oxidase synthase; nitric oxide synthase; nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 1, neuronal; nitric oxide synthase, brain; nNOS; N-NOS; nNOS; NOS type I; NO; NOS; NOS Type 1; NOS type I; Nos1; Nos-1; NOS-I; Peptidyl-cysteine S-nitrosylase NOS1; Phospho-bNOS; Phospho-Brain NOS | |
| Mouse | |
| Protein G | |
| RUO | |
| 18125, 24598 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction