missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NNT Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68194-1-IG-150UL
This item is not returnable.
View return policy
Description
The NNT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NNT. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The NNT antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| NNT | |
| Monoclonal | |
| 500 μg/mL | |
| MGC126502; MGC126503 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 114 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 1F5H6 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 23530 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction