missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: ANLLKTVVTG CSCPLLSNLG SCKGLRVKKD FLRTFYTHQE LWCKAPVKPG IPYKQLTVGV PKEIFQNEKR VALSPAGVQN LVKQGFNVVV ESGAGEASKF SDDHYRVAGA QIQGAKEVLA SDLVVKVRAP MVNPTLGVHE ADLLKTSGT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
This gene encodes an integral protein of the inner mitochondrial membrane. The enzyme couples hydride transfer between NAD(H) and NADP(+) to proton translocation across the inner mitochondrial membrane. Under most physiological conditions, the enzyme uses energy from the mitochondrial proton gradient to produce high concentrations of NADPH. The resulting NADPH is used for biosynthesis and in free radical detoxification.
Specifications
Specifications
| Antigen | NNT |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NNT |
| Gene Accession No. | Q13423 |
| Gene Alias | 4930423F13Rik; AI323702; BB168308; GCCD4; NAD(P) transhydrogenase, mitochondrial; nicotinamide nucleotide transhydrogenase; Nicotinamide nucleotide transhydrogenase (NAD(P)+ transhydrogenase); NNT; Pyridine nucleotide transhydrogenase; pyridine nucleotide transhydrogenase precursor (EC 1.6.1.1) |
| Gene Symbols | NNT |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
