missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NO66 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | NO66 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254125
|
Novus Biologicals
NBP2-58770 |
100 μL |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638178
|
Novus Biologicals
NBP2-58770-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NO66 Polyclonal specifically detects NO66 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NO66 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 79697 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RVVETSALLCTAQHLAAVQSSGAPATASGPQVDNTGGEPAWDSPLRRVLAELNRIPSSRRRAARLFEWLIAPMPPDHFYRRLWEREAVLVRRQDHTYYQGLFSTADLDSMLRNEEVQFGQHLDAARYINGRR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| chromosome 14 open reading frame 169, EC 1.14.11.27, FLJ21802, hsNO66, MAPJD, MYC-associated protein with JmjC domain, NO66lysine-specific demethylase NO66, Nucleolar protein 66 | |
| C14ORF169 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title