Learn More
Novus Biologicals™ NO66 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29177
This item is not returnable.
View return policy
Description
NO66 Polyclonal antibody specifically detects NO66 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NO66 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79697 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| chromosome 14 open reading frame 169, EC 1.14.11.27, FLJ21802, hsNO66, MAPJD, MYC-associated protein with JmjC domain, NO66lysine-specific demethylase NO66, Nucleolar protein 66 | |
| between 50 and 100 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |