missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Monoclonal Antibody (OTI4H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808806S
This item is not returnable.
View return policy
Description
NOB1 Monoclonal antibody specifically detects NOB1 in Human samples. It is validated for Western BlotSpecifications
| NOB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WZ19 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 253980 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4H3 | |
| Unconjugated | |
| KCTD13 | |
| 1700021I09Rik; 20S-pre-rRNA D-site endonuclease NOB1; adenocarcinoma antigen recognized by T lymphocytes 4; ART4; ART-4; MST158; MSTP158; nin one binding protein; NIN1/PSMD8 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog (S. cerevisiae); NIN1-binding protein; Nob1; Nob1p; phosphorylation regulatory protein HP-10; Pre-rRNA-processing endonuclease NOB1; Protein ART-4; PSMD8 binding protein 1; PSMD8BP1; RNA-binding protein NOB1; rRNA-binding endoribonuclease; YOR056C; YOR29-07 | |
| Full length human recombinant protein of human KCTD13 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction