missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Monoclonal Antibody (OTI4H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808806
This item is not returnable.
View return policy
Description
In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.Specifications
| NOB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9ULX3 | |
| NOB1 | |
| Human recombit protein fragment corresponding to amino acids 2-234 of human NOB1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4H3 | |
| Unconjugated | |
| NOB1 | |
| ART-4, MST158, MSTP158, NOB1P, PSMD8BP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 28987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction