missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOB1 Polyclonal Antibody

Product Code. 15729771 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15729771 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15729771 Supplier Invitrogen™ Supplier No. PA561967

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PSNIKQIQQE LEQCDVPEDV RVGCLTTDFA MQNVLLQMGL HVLAVNGMLI REARSYILRC HGCFKTTSDM SRVFCSHCGN KTLKKVSVTV SDDGTLHMHF SR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nob1
Gene Accession No. Q9ULX3
Gene Alias 1700021I09Rik; 20S-pre-rRNA D-site endonuclease NOB1; adenocarcinoma antigen recognized by T lymphocytes 4; ART4; ART-4; MST158; MSTP158; nin one binding protein; NIN1/PSMD8 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog (S. cerevisiae); NIN1-binding protein; Nob1; Nob1p; phosphorylation regulatory protein HP-10; Pre-rRNA-processing endonuclease NOB1; Protein ART-4; PSMD8 binding protein 1; PSMD8BP1; RNA-binding protein NOB1; rRNA-binding endoribonuclease; YOR056C; YOR29-07
Gene Symbols Nob1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOB1. Recombinant protein control fragment (Product #RP-98724).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.