missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nobiletin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Nobiletin is a poly-methoxylated flavone from the citrus peel that improves memory loss. Nobiletin is a retinoid acid receptor-related orphan receptors (RORs) agonist. Nobiletin can reduce reactive oxygen species (ROS) levels in differentiated C2C12 myotubes and has anti-inflammation and anti-cancer properties, including anti-angiogenesis, anti-proliferation, anti-metastasis and induced apoptosis.
Specifications
Specifications
| Chemical Name or Material | Nobiletin |
| CAS | 478-01-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H22O8 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (124.26 mM; Need ultrasonic) |
| SMILES | O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=C(OC)C(OC)=C(OC)C(OC)=C13 |
| Molecular Weight (g/mol) | 402.39 |
| Formula Weight | 402.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction