missing translation for 'onlineSavingsMsg'
Learn More

NOBOX Antibody (OTI4C5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73028APC

Product Code. 30024944

  • ONLINE EXCLUSIVE PRICE
    € 774.00
    € 731.85 / 0.10mL
    Save € 42.15
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NOBOX Monoclonal antibody specifically detects NOBOX in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NOBOX
Monoclonal
APC
PBS
Mouse
Immunogen affinity purified
RUO
135935
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry
OTI4C5
Western Blot, Immunohistochemistry
NOBOX NOBOX oogenesis homeobox, OG2, OG-2, OG2X, POF5, TCAG_12042
Human recombinant protein fragment corresponding to amino acids 1-271 of human NOBOX(NP_001073882) produced in E.coli.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.