missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NOC2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29166
This item is not returnable.
View return policy
Description
NOC2L Polyclonal antibody specifically detects NOC2L in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| NOC2L | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| DKFZP564C186, FLJ35172, NET15, NET7, NIR, NOC2-like protein, novel INHAT (inhibitor of histone acetyltransferase) repressor, nucleolar complex associated 2 homolog (S. cerevisiae), nucleolar complex protein 2 homolog, Protein NOC2 homolog | |
| Between 699 and 749 | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication | |
| 26155 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction