missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Nociceptin

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Nociceptin is an endogenous ligand for the NOP opioid receptor (pKi = 8.6). Has no significant activity at μ, δ or κ opioid receptors (pKi < 6).
Specifications
Specifications
| Description | Nociceptin |
| Molecular Weight (g/mol) | 1809 |
| Product Type | Nociceptin |
| Quantity | 1 mg |
| Sequence | Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction