missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nocturnin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37856
This item is not returnable.
View return policy
Description
Nocturnin Polyclonal specifically detects Nocturnin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nocturnin | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9UK39 | |
| CCRN4L | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AVTHLKARTGWERFRSAQGCDLLQNLQNITQGAKIPLIVCGDFNAEPTEEVYKHFASSSLNLNSAYKLLSADGQSEPPYTTWKIRTSGECRHTLDYIWYSKHALNVRSALDLLTEEQIGPNRLPSFNYPSDHLSLVCDFSFT | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CCR4 carbon catabolite repression 4-like (S. cerevisiae), CCR4 protein homolog, CCR4-like (carbon catabolite repression 4, S.cerevisiae), CCR4LMGC142060, CCR4MGC4120817, MGC142054, MGC78549, NOC, nocturnin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25819 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction