missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nodal Monoclonal Antibody (209009)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.
Specifications
Specifications
| Antigen | Nodal |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 209009 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | NODAL |
| Gene Accession No. | P43021 |
| Gene Alias | early embryo mesoderm formation; HTX5; MGC138230; Nodal; nodal growth differentiation factor; nodal homolog; Tg.413d; transgenic lethal 413.d |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?