missing translation for 'onlineSavingsMsg'
Learn More

NODAL Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen NODAL
Concentration 200 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236511
View Documents
Proteintech
27430-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237671
View Documents
Proteintech
27430-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The NODAL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NODAL. This antibody recognizes human antigen. The NODAL antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.
Specifications

Specifications

NODAL
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Unconjugated
Rabbit
Human
MGC138230
IgG
Antigen affinity purification
Polyclonal Antibody
200 μg/mL
Polyclonal
Liquid
RUO
4838
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.