missing translation for 'onlineSavingsMsg'
Learn More

Noggin Antibody (OTI1H8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73030C

Product Code. 18163386

  • € 785.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Noggin
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
NOG
Recombinant protein fragment corresponding to amino acids 28-232 of human Noggin (NP_005441) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1H8
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Mouse
Affinity Purified
Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
9241.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.