missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noggin Antibody (OTI1H8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73030H
This item is not returnable.
View return policy
Description
Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Noggin | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NOG | |
| Recombinant protein fragment corresponding to amino acids 28-232 of human Noggin (NP_005441) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1H8 | |
| Western Blot, Immunohistochemistry | |
| noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 9241.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction