missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Noggin Monoclonal Antibody (A11)

Mouse Monoclonal Antibody
€ 214.00 - € 518.00
Specifications
| Antigen | Noggin |
|---|---|
| Clone | A11 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17827002
![]() |
Invitrogen™
MA541654 |
200 μL |
€ 518.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17684166
|
Invitrogen™
MA543714 |
20 μL |
€ 214.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Noggin Monoclonal Antibody for Western Blot, IHC (P)
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.Specifications
| Noggin | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13253, Q62809 | |
| 25495, 9241 | |
| Recombinant protein Noggin. The antigen corresponds to amino acid range 28-232 of the target protein. | |
| Antibody |
| A11 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human, Rat | |
| NOG | |
| NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A | |
| NOG | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title