missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Noggin Monoclonal Antibody (A9)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA541655

 View more versions of this product

Product Code. 17806802

  • € 504.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Noggin Monoclonal Antibody for Western Blot, IHC (P)

Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Noggin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P97466, Q13253, Q62809
NOG
Recombinant protein Noggin. The antigen corresponds to amino acid range 28-232 of the target protein.
200 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot, Western Blot
A9
Unconjugated
NOG
NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A
Mouse
Protein A/G
RUO
18121, 25495, 9241
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.