missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo Antibody (atinumab), Alexa Fluor™ 405, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27996AF405
This item is not returnable.
View return policy
Description
Nogo Monoclonal antibody specifically detects Nogo in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Nogo | |
| Monoclonal | |
| Alexa Fluor 405 | |
| ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x | |
| RTN4/ NOGO | |
| 0.1 mL | |
| Apoptosis | |
| 57142 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| atinumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction