missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo Receptor/NgR Antibody (MM0493-6H43), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11823AF532
This item is not returnable.
View return policy
Description
Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human samples. It is validated for Western Blot.
Specifications
| Nogo Receptor/NgR | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RTN4R | |
| Human recombinant Nogo Receptor/NgR (extracellular domain) | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot | |
| MM0493-6H43 | |
| Western Blot | |
| NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
| Mouse | |
| 0.1 mL | |
| 65078 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction