missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18907R
This item is not returnable.
View return policy
Description
NOL1 Polyclonal antibody specifically detects NOL1 in Human samples. It is validated for Western Blot
Specifications
| NOL1 | |
| Polyclonal | |
| Western Blot | |
| EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2 | |
| The immunogen recognized by this antibody maps to a region between residue 762 and 812 of human nucleolar protein 1 using the numbering given in entry AAH00656.1 (GeneID 4839). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4839 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction