missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL10 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47269AF594
This item is not returnable.
View return policy
Description
NOL10 Polyclonal antibody specifically detects NOL10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NOL10 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ13938, FLJ14075, H_NH0074G24.1, nucleolar protein 10, polyglutamine binding protein 5, PQBP5 | |
| The immunogen for this product maps to a region between residue 638 and 688 of human Nucleolar Protein 10 using the numbering given in entry NP_079170.2 (GeneID 79954). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79954 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction