missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOL10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17711984
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17711984 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17711984 Supplier Invitrogen™ Supplier No. PA5120581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, Mouse thymus, Mouse testis, Mouse spleen. Immunogen sequence: KNSGKIFTSL EPEHDLNDVC LYPNSGMLLT ANETPKMGIY YIPVLGPAPR WCSFLDNLTE ELEENPESTV YDDYKFVTKK DLENLGLTHL IGSPFLRAYM H.

NOL10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NOL10
Gene Accession No. Q5RJG1, Q9BSC4
Gene Alias Gm67; H_NH0074G24.1; NOL10; Nucleolar protein 10; polyglutamine binding protein 5; PQBP5
Gene Symbols NOL10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-400 of human NOL10 (NP_079170.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217431, 79954
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.