missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL12 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46174B
This item is not returnable.
View return policy
Description
NOL12 Polyclonal antibody specifically detects NOL12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NOL12 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| dJ37E16.7, FLJ34609, MGC3731, Nop25, nucleolar protein 12, RRP17 | |
| The immunogen for this product maps to a region between residue 163 and 213 of human Nucleolar Protein 12 using the numbering given in entry NP_077289.1 (GeneID 79159). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79159 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction