missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19560
This item is not returnable.
View return policy
Description
NOL12 Polyclonal specifically detects NOL12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NOL12 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| dJ37E16.7, FLJ34609, MGC3731, Nop25, nucleolar protein 12, RRP17 | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9UGY1 | |
| NOL12 | |
| Recombinant protein encompassing a sequence within the center region of human Nop25. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79159 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction