missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL3, Mouse anti-Human, Clone: 3A9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008996-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MGNAQERPSETIDRERKRLVETLQADSGLLLDALLARGVLTGPEYEALDALPDAERRVRRLLLLVQGKGEAACQELLRCAQRTAGAPDPAWDWQHVGPGYRDRSYDPPCPGHWTPEAPGSGTTCPGLPRASDPDEAGGPEGSEAVQSGTPEEPEPELEAEASKEAEPEPEPEPELEPEAEAEPEPELEPEPDPEPEPDFEERDESEDSSpecifications
| NOL3 | |
| Monoclonal | |
| Unconjugated | |
| nucleolar protein 3 (apoptosis repressor with CARD domain) | |
| ARC/CARD2/MYC/MYP/NOP/NOP30 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3A9 | |
| In 1x PBS, pH 7.4 | |
| NM_003946.3 | |
| NOL3 | |
| NOL3 (NP_003937.1, 1 a.a. ∼ 208 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8996 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction