missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOL7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556079
This item is not returnable.
View return policy
Description
Immunogen sequence: EPLEEDEEGD DEFDDEAPEE LTFASAQAEA REEERRVRET VRRDKTLLKE KRKRREELFI EQKKRKL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
NOL7 is encoded by this gene localizes to the nucleolus, where it maintains nucleolar structure and cell growth rates. The encoded protein also functions as a tumor suppressor and regulator of angiogenesis. The RB tumor suppressor gene recruits transcription factors to this gene and positively regulates its expression. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| NOL7 | |
| Polyclonal | |
| Unconjugated | |
| NOL7 | |
| 2210008F15Rik; 5730556I21Rik; C6orf90; dJ223E5.2; MNCb-1192; NOL7; Nol7-ps1; NOP27; nucleolar protein 7; nucleolar protein 7, 27kDa; nucleolar protein 7, pseudogene 1; nucleolar protein of 27 kDa; polyglutamine binding protein 3; PQBP3; RARG-1; retinoic acid repressible protein; RGD1565934 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UMY1 | |
| NOL7 | |
| Recombinant protein corresponding to Human NOL7. Recombinant protein control fragment (Product #RP-94875). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction