missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOLA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93225-0.02ml
This item is not returnable.
View return policy
Description
NOLA1 Polyclonal antibody specifically detects NOLA1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| NOLA1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| GAR1 ribonucleoprotein homolog (yeast), H/ACA ribonucleoprotein complex subunit 1, NOLA1member 1 (H/ACA small nucleolar RNPs), Nucleolar protein family A member 1, snoRNP protein GAR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-170 of human GAR1 (NP_061856.1). KGQDQGPPERVVLLGEFLHPCEDDIVCKCTTDENKVPYFNAPVYLENKEQIGKVDEIFGQLRDFYFSVKLSENMKASSFKKLQKFYIDPYKLLPLQRFLPRPPGEKGPPRG | |
| 0.02 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54433 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction